ENA413170706 Enamine

1-(1-methyl-1H-imidazol-2-yl)-1-(oxan-4-yl)methanamine

  • MDL number MFCD21188343
  • Cas Number : 1482275-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153333

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153333
Availability details for
Quantity
  • Availability
  • Add to Cart