ENA413170709 Enamine

(1-methyl-2,3-dihydro-1H-indol-5-yl)methanamine dihydrochloride

  • MDL number MFCD27500829
  • Cas Number : 1607300-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153339

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153339
Availability details for
Quantity
  • Availability
  • Add to Cart