ENA413170712 Enamine

5-bromo-3-(trifluoromethyl)-1,2-benzoxazole

  • MDL number MFCD15526913
  • Cas Number : 1260790-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153343

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153343
Availability details for
Quantity
  • Availability
  • Add to Cart