ENA413170714 Enamine

4-[(2-carbamoylethyl)amino]benzoic acid

  • MDL number MFCD16692395
  • Cas Number : 1249907-84-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153347

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153347
Availability details for
Quantity
  • Availability
  • Add to Cart