ENA413170717 Enamine

1-(2-fluoro-6-methoxyphenyl)piperazine

  • MDL number MFCD11872779
  • Cas Number : 113028-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153356

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153356
Availability details for
Quantity
  • Availability
  • Add to Cart