ENA413170726 Enamine

4-N-(4-methylphenyl)pyridine-3,4-diamine

  • MDL number MFCD11202092
  • Cas Number : 1038315-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153436

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153436
Availability details for
Quantity
  • Availability
  • Add to Cart