ENA413170728 Enamine

N-(4-methylphenyl)-3-nitropyridin-4-amine

  • MDL number MFCD27500833
  • Cas Number : 54696-69-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153438

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153438
Availability details for
Quantity
  • Availability
  • Add to Cart