ENA413170734 Enamine

1,3-diethyl 2-(cyclohex-2-en-1-yl)propanedioate

  • MDL number MFCD27500834
  • Cas Number : 6305-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153448

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153448
Availability details for
Quantity
  • Availability
  • Add to Cart