ENA413170743 Enamine

1-(2,2,2-trifluoroethyl)-1H-pyrazole-4-sulfonyl chloride

  • MDL number MFCD08696567
  • Cas Number : 1006487-19-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153475

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153475
Availability details for
Quantity
  • Availability
  • Add to Cart