ENA413170747 Enamine

methyl 4-(1-aminoethyl)piperidine-1-carboxylate hydrochloride

  • MDL number MFCD27500838
  • Cas Number : 1803610-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153489

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153489
Availability details for
Quantity
  • Availability
  • Add to Cart