ENA413170749 Enamine

2-[4-(1-aminoethyl)phenyl]-1$l^{6},2-thiazolidine-1,1-dione hydrochloride

  • MDL number MFCD27500840
  • Cas Number : 1607250-69-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153493

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153493
Availability details for
Quantity
  • Availability
  • Add to Cart