ENA413170764 Enamine

2-[(1-{[(9H-fluoren-9-yl)methoxy]carbonyl}piperidin-4-yl)oxy]acetic acid

  • MDL number MFCD18903920
  • Cas Number : 1341147-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153567

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153567
Availability details for
Quantity
  • Availability
  • Add to Cart