ENA413170767 Enamine

(4S)-6,8-difluoro-3,4-dihydro-2H-1-benzopyran-4-ol

  • MDL number MFCD15478371
  • Cas Number : 1270301-86-4
  • Molecular Weight : 186.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-153601

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153601
Availability details for
Quantity
  • Availability
  • Add to Cart