ENA413170770 Enamine

3-tert-butyl-1-(propan-2-yl)thiourea

  • MDL number MFCD03427358
  • Cas Number : 52599-24-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153613

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153613
Availability details for
Quantity
  • Availability
  • Add to Cart