ENA413170772 Enamine

2-(4-tert-butyl-1,2,3,6-tetrahydropyridin-1-yl)ethan-1-amine dihydrochloride

  • MDL number MFCD27500850
  • Cas Number : 1803582-70-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153616

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153616
Availability details for
Quantity
  • Availability
  • Add to Cart