ENA413170779 Enamine

methyl 2-methylpyrrolidine-2-carboxylate

  • MDL number MFCD19217446
  • Cas Number : 51098-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153625

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-153625
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart