ENA413170785 Enamine

2-(aminomethyl)-5-methyl-[1,2,4]triazolo[1,5-a]pyrimidin-7-ol

  • MDL number MFCD20724762
  • Cas Number : 150190-92-4
  • Molecular Weight : 179.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-153635

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153635
Availability details for
Quantity
  • Availability
  • Add to Cart