ENA413170786 Enamine

N-[(3-{[(diaminomethylidene)amino]methyl}phenyl)methyl]guanidine dihydrochloride

  • MDL number MFCD27500855
  • Cas Number : 154474-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-153638

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153638
Availability details for
Quantity
  • Availability
  • Add to Cart