ENA413170792 Enamine

methyl 2-amino-3-fluorobenzoate

  • MDL number MFCD11870179
  • Cas Number : 144851-82-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153703
Availability details for
Quantity
  • Availability
  • Add to Cart