ENA413170794 Enamine

(4R)-6,8-difluoro-3,4-dihydro-2H-1-benzopyran-4-amine hydrochloride

  • MDL number MFCD12913731
  • Cas Number : 911826-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153729

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153729
Availability details for
Quantity
  • Availability
  • Add to Cart