ENA413170798 Enamine

2-(prop-2-en-1-ylsulfanyl)benzoic acid

  • MDL number MFCD07368287
  • Cas Number : 17839-66-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153736

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153736
Availability details for
Quantity
  • Availability
  • Add to Cart