ENA413170801 Enamine

1-{[(carbamoylamino)carbonyl]amino}formamide

  • MDL number MFCD00025439
  • Cas Number : 556-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153744

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153744
Availability details for
Quantity
  • Availability
  • Add to Cart