ENA413170804 Enamine

3-(5-methylthiophen-3-yl)pyrrolidine

  • MDL number MFCD17484117
  • Cas Number : 1260842-14-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153748

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153748
Availability details for
Quantity
  • Availability
  • Add to Cart