ENA413170812 Enamine

8-fluoroquinoline-3-carbaldehyde

  • MDL number MFCD18433599
  • Cas Number : 1258638-54-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153760

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153760
Availability details for
Quantity
  • Availability
  • Add to Cart