ENA413170813 Enamine

1-(8-fluoroquinolin-3-yl)methanamine

  • MDL number MFCD19285294
  • Cas Number : 1267242-11-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153761

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153761
Availability details for
Quantity
  • Availability
  • Add to Cart