ENA413170821 Enamine

1-(5-fluoro-2-nitrophenyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD17154090

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-151995

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-151995
Availability details for
Quantity
  • Availability
  • Add to Cart