ENA413170824 Enamine

8-fluoroquinoline-3-carbonitrile

  • MDL number MFCD27500875
  • Cas Number : 71083-52-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175-176
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153773

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153773
Availability details for
Quantity
  • Availability
  • Add to Cart