ENA413170832 Enamine

5-{[(tert-butoxy)carbonyl]amino}-3-methyl-1,2-thiazole-4-carboxylic acid

  • MDL number MFCD13159692
  • Cas Number : 1179765-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153790

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153790
Availability details for
Quantity
  • Availability
  • Add to Cart