ENA413170838 Enamine

tert-butyl (2R)-2-(cyanomethyl)pyrrolidine-1-carboxylate

  • MDL number MFCD11505915
  • Cas Number : 201039-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153803

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153803
Availability details for
Quantity
  • Availability
  • Add to Cart