ENA413170849 Enamine

tert-butyl (2S)-2-[(methanesulfonyloxy)methyl]pyrrolidine-1-carboxylate

  • MDL number MFCD27500883
  • Cas Number : 132482-09-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153847

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153847
Availability details for
Quantity
  • Availability
  • Add to Cart