ENA413170860 Enamine

4-methyl-2-{3-[1-(methylamino)ethyl]phenyl}-1$l^{6},2-thiazolidine-1,1-dione

  • MDL number MFCD24745328
  • Cas Number : 1512042-24-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153882

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153882
Availability details for
Quantity
  • Availability
  • Add to Cart