ENA413170862 Enamine

4-methyl-2-propyl-1,3-thiazol-5-amine

  • MDL number MFCD16669993
  • Cas Number : 1250631-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153886

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153886
Availability details for
Quantity
  • Availability
  • Add to Cart