ENA413170864 Enamine

N-tert-butyl-2-(methylamino)propanamide

  • MDL number MFCD11176746
  • Cas Number : 1132791-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153889

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153889
Availability details for
Quantity
  • Availability
  • Add to Cart