ENA413170865 Enamine

methyl 6-methoxy-2-oxo-2,3-dihydro-1H-1,3-benzodiazole-5-carboxylate

  • MDL number MFCD22562234
  • Cas Number : 1388063-51-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153894

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153894
Availability details for
Quantity
  • Availability
  • Add to Cart