ENA413170869 Enamine

6,8-dimethylisoquinoline

  • MDL number MFCD18448351
  • Cas Number : 67878-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153928

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153928
Availability details for
Quantity
  • Availability
  • Add to Cart