ENA413170870 Enamine

2-(phenylamino)-4-(trifluoromethyl)pyrimidine-5-carboxylic acid

  • MDL number MFCD16657327
  • Cas Number : 666260-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153931

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153931
Availability details for
Quantity
  • Availability
  • Add to Cart