ENA413170872 Enamine

5-bromo-2-methoxy-6-methylpyridine-3-carbonitrile

  • MDL number MFCD27500891
  • Cas Number : 88818-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153941

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153941
Availability details for
Quantity
  • Availability
  • Add to Cart