ENA413170873 Enamine

5-bromo-2-methoxy-6-methylpyridine-3-carboxylic acid

  • MDL number MFCD22033941
  • Cas Number : 1369354-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153943

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153943
Availability details for
Quantity
  • Availability
  • Add to Cart