ENA413170884 Enamine

1-(2-oxo-2-phenylethyl)-1,2-dihydropyrimidin-2-one

  • MDL number MFCD16154112
  • Cas Number : 933053-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154008

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154008
Availability details for
Quantity
  • Availability
  • Add to Cart