ENA413170891 Enamine

methyl 2-methyl-2-(piperazin-1-yl)propanoate

  • MDL number MFCD12818035
  • Cas Number : 1182949-28-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154045

Safety Information

msds https://www.enaminestore.com/catalog/EN300-154045
Availability details for
Quantity
  • Availability
  • Add to Cart