ENA413170899 Enamine

1,3-dioxo-2,3-dihydro-1H-isoindole-4-carboxylic acid

  • MDL number MFCD20639778
  • Cas Number : 776-22-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154064

Safety Information

msds https://www.enaminestore.com/catalog/EN300-154064
Availability details for
Quantity
  • Availability
  • Add to Cart