ENA413170912 Enamine

tert-butyl 3-formyl-3-methylpyrrolidine-1-carboxylate

  • MDL number MFCD14582389
  • Cas Number : 1205748-72-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154094

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154094
Availability details for
Quantity
  • Availability
  • Add to Cart