ENA413170917 Enamine

N-[(1H-imidazol-4-yl)methyl]aniline

  • MDL number MFCD27500903
  • Cas Number : 32563-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154105

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154105
Availability details for
Quantity
  • Availability
  • Add to Cart