ENA413170919 Enamine

methyl 1-(aminomethyl)cyclopropane-1-carboxylate hydrochloride

  • MDL number MFCD10758090
  • Cas Number : 1170782-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154111

Safety Information

msds https://www.enaminestore.com/catalog/EN300-154111
Availability details for
Quantity
  • Availability
  • Add to Cart