ENA413170920 Enamine

5-(aminomethyl)-1,3,4-oxadiazol-2-amine dihydrochloride

  • MDL number MFCD27500905
  • Cas Number : 89066-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154113

Safety Information

msds https://www.enaminestore.com/catalog/EN300-154113
Availability details for
Quantity
  • Availability
  • Add to Cart