ENA413170923 Enamine

2-methylprop-2-ene-1-sulfonamide

  • MDL number MFCD21820877
  • Cas Number : 39222-55-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154128

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154128
Availability details for
Quantity
  • Availability
  • Add to Cart