ENA413170928 Enamine

[3-methyl-1-(methylamino)cyclopentyl]methanol

  • MDL number MFCD20315284
  • Cas Number : 1339535-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154160

Safety Information

msds https://www.enaminestore.com/catalog/EN300-154160
Availability details for
Quantity
  • Availability
  • Add to Cart