ENA413170939 Enamine

tert-butyl 3-(bromomethyl)pyrrolidine-1-carboxylate

  • MDL number MFCD06410515
  • Cas Number : 305329-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154209

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-154209
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart