ENA413170972 Enamine

tert-butyl (2R)-2-(2-aminoethyl)pyrrolidine-1-carboxylate

  • MDL number MFCD04115290
  • Cas Number : 550378-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154302

Safety Information

msds https://www.enaminestore.com/catalog/EN300-154302
Availability details for
Quantity
  • Availability
  • Add to Cart