ENA413170985 Enamine

ethyl 3-amino-3-cyclohexylbutanoate

  • MDL number MFCD23782385
  • Cas Number : 1512113-65-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154360

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154360
Availability details for
Quantity
  • Availability
  • Add to Cart